Assay Details
Target Gene Details
Entrez Gene ID: | 67412 |
Gene Name: | SOGA family member 3 |
Gene Aliases: |
6330407J23Rik |
Location: |
Chr.10:29143274-29199630 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Soga3 | NM_026138.2 | 3 | 1696 | NP_080414.2 |
| AK018139.1 | 2 | 226 | BAB31090.1 | |
| AK030682.1 | 3 | 1569 | ||
| AK044134.1 | 2 | 551 | BAC31794.2 | |
| AK163577.1 | 2 | 196 | ||
| BC066079.1 | 3 | 1853 | AAH66079.1 |

Genomic Map