Assay Details
Target Gene Details
Entrez Gene ID: | 28194 |
Gene Name: | apolipoprotein N |
Gene Aliases: |
AI195275, D10Ucla2 |
Location: |
Chr.10:128254131-128255901 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apon | NM_133996.3 | NP_598757.1 | ||
| AK083289.1 | BAC38845.1 | |||
| BY096905.1 |
More Information
Set Membership: |
|

Genomic Map