Assay Details
Target Gene Details
Entrez Gene ID: | 71564 |
Gene Name: | IZUMO family member 4 |
Gene Aliases: |
9030607L17Rik, AI550407 |
Location: |
Chr.10:80702199-80705382 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Izumo4 | NM_027829.3 | NP_082105.3 | ||
| AK018541.1 | 4 | 369 | BAB31263.1 | |
| BC057953.1 | AAH57953.1 | |||
| BF682661.1 | ||||
| CB055593.1 |
More Information
Set Membership: |
|

Genomic Map