Assay Details
Target Gene Details
Entrez Gene ID: | 216001 |
Gene Name: | mitochondrial calcium uptake 1 |
Gene Aliases: |
C730016L05Rik, Calc, Cbara1 |
Location: |
Chr.10:59702482-59864134 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Micu1 | NM_001291442.1 | 9 | 1152 | NP_001278371.1 |
| NM_001291443.1 | 10 | 1146 | NP_001278372.1 | |
| NM_144822.3 | 9 | 1134 | NP_659071.1 | |
| AK046049.1 | 9 | 1117 | BAC32581.1 | |
| AK049521.1 | 9 | 1130 | BAC33792.1 | |
| AK050113.1 | 9 | 1121 | BAC34073.1 | |
| AK052485.1 | 9 | 1082 | BAC35013.1 | |
| AK077605.1 | 9 | 1084 | BAC36895.1 | |
| AK085553.1 | 9 | 1053 | ||
| AK185405.1 | 2 | 174 | ||
| AK205758.1 | ||||
| AK212389.1 | ||||
| AK212821.1 | ||||
| BC018320.1 | 9 | 1075 | AAH18320.1 | |
| BC023022.1 | 10 | 1081 | AAH23022.1 | |
| BC023042.1 | 8 | 1018 | AAH23042.1 | |
| BC026566.1 | 9 | 1085 | AAH26566.1 |
More Information
Set Membership: |
|

Genomic Map