Assay Details
Target Gene Details
Entrez Gene ID: | 70615 |
Gene Name: | ankyrin repeat domain 24 |
Gene Aliases: |
4631433D01Rik, 5730519E19Rik, D10Bur2e |
Location: |
Chr.10:81627795-81647612 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd24 | NM_027480.3 | NP_081756.4 | ||
| AK043638.1 | ||||
| AK173316.1 | BAD32594.1 | |||
| BC048236.1 | ||||
| CJ177454.1 | ||||
| CJ183294.1 |

Genomic Map