Assay Details
Target Gene Details
Entrez Gene ID: | 237615 |
Gene Name: | ankyrin repeat domain 52 |
Gene Aliases: |
6430544C07, G431002C21Rik |
Location: |
Chr.10:128377124-128394006 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd52 | NM_172790.2 | NP_766378.1 | ||
| AK032429.1 | 15 | 1735 | BAC27865.1 | |
| AK035533.1 | 15 | 1720 | ||
| AK090124.1 | BAC41105.1 | |||
| AK144017.1 | 15 | 1742 | BAE25660.1 | |
| BC117907.1 | ||||
| BC117908.1 |
More Information
Set Membership: |
|

Genomic Map