Assay Details
Target Gene Details
Entrez Gene ID: | 14533 |
Gene Name: | biogenesis of lysosomal organelles complex-1, subunit 1 |
Gene Aliases: |
AI839753, BLOS1, Gcn5l1 |
Location: |
Chr.10:128919220-128923547 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bloc1s1 | NM_015740.3 | NP_056555.2 | ||
| AK003035.1 | BAB22523.1 | |||
| BC034662.1 | AAH34662.1 |
More Information
Set Membership: |
|

Genomic Map