Assay Details
Target Gene Details
Entrez Gene ID: | 215999 |
Gene Name: | mitochondrial calcium uniporter |
Gene Aliases: |
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64 |
Location: |
Chr.10:59446984-59616692 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcu | NM_001033259.4 | NP_001028431.2 | ||
| AK144727.1 | BAE26033.1 |

Genomic Map