Assay Details
Target Gene Details
Entrez Gene ID: | 12566 |
Gene Name: | cyclin-dependent kinase 2 |
Gene Aliases: |
A630093N05Rik |
Location: |
Chr.10:128697938-128705051 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk2 | NM_016756.4 | 4 | 625 | NP_058036.1 |
| NM_183417.3 | 4 | 625 | NP_904326.1 | |
| AJ223732.1 | 4 | 527 | CAA11533.1 | |
| AK036589.1 | 1 | 2507 | ||
| AK042462.1 | 3 | 1286 | ||
| AK146845.1 | 4 | 605 | BAE27476.1 | |
| AK148017.1 | 4 | 564 | BAE28290.1 | |
| AK151979.1 | 4 | 575 | BAE30846.1 | |
| AK152922.1 | 4 | 578 | BAE31597.1 | |
| AK153983.1 | 4 | 577 | BAE32294.1 | |
| AK154997.1 | 4 | 577 | BAE32983.1 | |
| AK179714.1 | 3 | 297 | ||
| AK181235.1 | 3 | 296 | ||
| AK184593.1 | 3 | 294 | ||
| AK186022.1 | 3 | 296 | ||
| AK186779.1 | ||||
| AK187629.1 | 4 | 361 | ||
| AK189612.1 | 4 | 361 | ||
| AK200296.1 | 1 | 58 | ||
| AK210959.1 | 3 | 296 | ||
| AK219393.1 | 3 | 296 | ||
| BC005654.1 | 4 | 586 | AAH05654.1 | |
| U63337.1 | 4 | 553 | AAB37128.1 |
More Information
Set Membership: |
|

Genomic Map