Assay Details
Target Gene Details
Entrez Gene ID: | 12566 |
Gene Name: | cyclin-dependent kinase 2 |
Gene Aliases: |
A630093N05Rik |
Location: |
Chr.10:128697938-128705051 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk2 | NM_016756.4 | 6 | 955 | NP_058036.1 |
| NM_183417.3 | 7 | 1099 | NP_904326.1 | |
| AJ223732.1 | 7 | 1001 | CAA11533.1 | |
| AK146845.1 | 7 | 1079 | BAE27476.1 | |
| AK148017.1 | 7 | 1038 | BAE28290.1 | |
| AK151979.1 | 7 | 1049 | BAE30846.1 | |
| AK152922.1 | 6 | 908 | BAE31597.1 | |
| AK153983.1 | 7 | 1051 | BAE32294.1 | |
| AK154997.1 | 7 | 1051 | BAE32983.1 | |
| AK178832.1 | 1 | 92 | ||
| AK179397.1 | 1 | 92 | ||
| AK183240.1 | 1 | 62 | ||
| BC005654.1 | 7 | 1060 | AAH05654.1 | |
| U63337.1 | 6 | 883 | AAB37128.1 |
More Information
Set Membership: |
|

Genomic Map