Assay Details
Target Gene Details
Entrez Gene ID: | 70312 |
Gene Name: | cactin, spliceosome C complex subunit |
Gene Aliases: |
2510012J08Rik |
Location: |
Chr.10:81321103-81326251 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cactin | NM_027381.2 | NP_081657.1 | ||
| AY934504.1 | ||||
| BC150986.1 | ||||
| BC151180.1 |
More Information
Set Membership: |
|

Genomic Map