Assay Details
Target Gene Details
Entrez Gene ID: | 270757 |
Gene Name: | BPI fold containing family C |
Gene Aliases: |
4732454E24, Bpil2 |
Location: |
Chr.10:85959340-86015898 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bpifc | NM_177772.4 | NP_808440.2 | ||
| AK028764.1 | BAC26108.1 | |||
| AK132518.1 | BAE21214.1 | |||
| BC112376.1 | AAI12377.1 | |||
| BY142313.1 |

Genomic Map