Assay Details
Target Gene Details
Entrez Gene ID: | 13608 |
Gene Name: | ectodysplasin-A receptor |
Gene Aliases: |
ED1R, ED3, ED5, EDA-A1R, EDA3, dl |
Location: |
Chr.10:58600780-58675696 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Edar | NM_010100.3 | NP_034230.1 | ||
| AF160502.1 | AAD50425.1 | |||
| AK004576.2 | BAB23385.1 | |||
| BC068315.1 | AAH68315.1 |
More Information
Set Membership: |
|

Genomic Map