Assay Details
Target Gene Details
Entrez Gene ID: | 69718 |
Gene Name: | inositol polyphosphate multikinase |
Gene Aliases: |
2410017C19Rik, AA408208, Impk |
Location: |
Chr.10:71347765-71385885 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ipmk | NM_027184.1 | NP_081460.1 | ||
| AK010523.1 | BAB27004.1 | |||
| AK036084.1 | BAC29300.1 | |||
| AK036978.1 | BAC29655.1 | |||
| AK172246.1 | ||||
| BC052463.1 | AAH52463.1 |
More Information
Set Membership: |
|

Genomic Map