Assay Details
Target Gene Details
Entrez Gene ID: | 69917 |
Gene Name: | nucleic acid binding protein 2 |
Gene Aliases: |
2610036N15Rik, Obfc2b |
Location: |
Chr.10:128401394-128411487 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nabp2 | NM_027257.1 | NP_081533.1 | ||
| BC026942.1 | AAH26942.1 | |||
| DQ158906.1 | AAZ85395.1 |
More Information
Set Membership: |
|

Genomic Map