Assay Details
Target Gene Details
Entrez Gene ID: | 216238 |
Gene Name: | early endosome antigen 1 |
Gene Aliases: |
A430109M19Rik, B230358H09Rik, ZFYVE2 |
Location: |
Chr.10:95940660-96045518 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eea1 | NM_001001932.3 | NP_001001932.1 | ||
| AK046231.1 | BAC32647.1 | |||
| BC075637.1 | AAH75637.1 |
More Information
Set Membership: |
|

Genomic Map