Assay Details
Target Gene Details
Entrez Gene ID: | 237397 |
Gene Name: | C2 calcium-dependent domain containing 4C |
Gene Aliases: |
4932409I22Rik, Fam148c, Gm238 |
Location: |
Chr.10:79606854-79614025 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C2cd4c | NM_001168624.1 | 1 | 63 | NP_001162095.1 |
| NM_198614.3 | 1 | 56 | NP_941016.2 | |
| AK029946.1 | 1 | 64 | BAC26694.1 | |
| BC089008.1 | 1 | 38 | AAH89008.1 | |
| BY268666.1 | 1 | 57 |

Genomic Map