Assay Details
Target Gene Details
Entrez Gene ID: | 68114 |
Gene Name: | melanoma associated antigen (mutated) 1 |
Gene Aliases: |
2610019J09, 9430059D04Rik, UBE-1C2 |
Location: |
Chr.10:80226434-80245144 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mum1 | NM_023431.5 | NP_075920.4 | ||
| AK011465.1 | BAB27636.1 | |||
| AK078792.1 | BAC37397.1 | |||
| AK147490.1 | 2 | 4020 | ||
| AK154610.1 | ||||
| AK158750.1 | BAE34640.1 | |||
| AK159972.1 | ||||
| AK170459.1 | BAE41811.1 | |||
| AK178196.1 | ||||
| AK186340.1 | ||||
| AK192032.1 | ||||
| AK219139.1 | ||||
| BC023031.1 | AAH23031.1 | |||
| BC069883.1 | AAH69883.1 | |||
| BC075617.1 | AAH75617.1 |
More Information
Set Membership: |
|

Genomic Map