Assay Details
Target Gene Details
Entrez Gene ID: | 211329 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
9030406N13Rik |
Location: |
Chr.10:30645582-30805977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa7 | NM_172495.5 | NP_766083.3 | ||
| AK128935.1 | BAC87660.1 | |||
| BC076623.1 | AAH76623.1 | |||
| BC141150.1 |

Genomic Map