Assay Details
Target Gene Details
Entrez Gene ID: | 211329 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
9030406N13Rik |
Location: |
Chr.10:30645582-30805977 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa7 | NM_001111267.2 | 3 | 439 | NP_001104737.1 |
| NM_172495.5 | 13 | 2790 | NP_766083.3 | |
| AK033503.1 | 3 | 353 | BAC28325.1 | |
| AK042369.1 | 3 | 409 | BAC31241.1 | |
| AK128935.1 | 12 | 2613 | BAC87660.1 | |
| AK165098.1 | 3 | 408 | ||
| AK171679.1 | 3 | 357 | BAE42607.1 | |
| BC049997.1 | 3 | 389 | AAH49997.1 | |
| BC058572.1 | 3 | 361 | AAH58572.1 | |
| BC076623.1 | 11 | 2396 | AAH76623.1 | |
| BC141150.1 | 12 | 2491 | ||
| EF602028.1 | 3 | 439 |

Genomic Map