Assay Details
Target Gene Details
Entrez Gene ID: | 211329 |
Gene Name: | nuclear receptor coactivator 7 |
Gene Aliases: |
9030406N13Rik |
Location: |
Chr.10:30645582-30805977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncoa7 | NM_001111267.2 | NP_001104737.1 | ||
| NM_172495.5 | NP_766083.3 | |||
| AK033503.1 | BAC28325.1 | |||
| AK042369.1 | 5 | 705 | BAC31241.1 | |
| AK128935.1 | BAC87660.1 | |||
| AK165098.1 | 4 | 1108 | ||
| AK171679.1 | BAE42607.1 | |||
| BC049997.1 | AAH49997.1 | |||
| BC058572.1 | AAH58572.1 | |||
| BC076623.1 | AAH76623.1 | |||
| BC141150.1 | ||||
| EF602028.1 |

Genomic Map