Assay Details
Target Gene Details
Entrez Gene ID: | 75122 |
Gene Name: | zinc finger, C2HC-type containing 1B |
Gene Aliases: |
4930519B02Rik, Fam164b |
Location: |
Chr.10:13143095-13178038 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zc2hc1b | NM_029172.1 | NP_083448.1 | ||
| AK015837.1 | BAB29995.1 | |||
| BC137685.1 | ||||
| BC144830.1 |
More Information
Set Membership: |
|

Genomic Map