Assay Details
Target Gene Details
Entrez Gene ID: | 68114 |
Gene Name: | melanoma associated antigen (mutated) 1 |
Gene Aliases: |
2610019J09, 9430059D04Rik, UBE-1C2 |
Location: |
Chr.10:80226434-80245144 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mum1 | NM_023431.5 | NP_075920.4 | ||
| AK078792.1 | BAC37397.1 | |||
| AK154610.1 | ||||
| AK158750.1 | BAE34640.1 | |||
| AK159972.1 | 3 | 1163 | ||
| AK170459.1 | BAE41811.1 | |||
| AK180155.1 | ||||
| AK205449.1 | ||||
| AK219009.1 | ||||
| AK220121.1 | ||||
| BC075617.1 | AAH75617.1 |
More Information
Set Membership: |
|

Genomic Map