Assay Details
Target Gene Details
Entrez Gene ID: | 13637 |
Gene Name: | ephrin A2 |
Gene Aliases: |
CEK7L, Elf1, Epl6, Eplg6, Lerk6 |
Location: |
Chr.10:80179482-80190010 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efna2 | NM_007909.3 | NP_031935.3 | ||
| AK140544.1 | BAE24419.1 | |||
| BC048697.1 | AAH48697.1 | |||
| U14752.1 | AAA68520.1 | |||
| U14941.1 | AAA53636.1 |
More Information
Set Membership: |
|

Genomic Map