Assay Details
Target Gene Details
Entrez Gene ID: | 75906 |
Gene Name: | family with sequence similarity 184, member A |
Gene Aliases: |
3110012E06Rik, 4930438C08Rik, 4930589M24Rik |
Location: |
Chr.10:53633128-53751231 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam184a | NM_001081428.1 | 4 | 1277 | NP_001074897.1 |

Genomic Map