Assay Details
Target Gene Details
Entrez Gene ID: | 237436 |
Gene Name: | growth arrest-specific 2 like 3 |
Gene Aliases: |
8430435B07Rik, Gm240 |
Location: |
Chr.10:89408822-89444374 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas2l3 | NM_001033331.2 | NP_001028503.2 | ||
| NM_001079876.1 | NP_001073345.1 | |||
| NM_001284344.1 | NP_001271273.1 | |||
| AK136054.1 | BAE22798.1 | |||
| AK164505.1 | BAE37814.1 | |||
| AK170634.1 | ||||
| AK170750.1 | BAE42001.1 |

Genomic Map