Assay Details
Target Gene Details
Entrez Gene ID: | 16971 |
Gene Name: | low density lipoprotein receptor-related protein 1 |
Gene Aliases: |
A2mr, AI316852, CD91, Lrp, b2b1554Clo |
Location: |
Chr.10:127538157-127621148 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 79 - Exon 79 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrp1 | NM_008512.2 | NP_032538.2 | ||
| AF367720.1 | AAL09566.1 | |||
| X67469.1 | CAA47817.1 |
More Information
Set Membership: |
|

Genomic Map