Assay Details
Target Gene Details
Entrez Gene ID: | 109272 |
Gene Name: | myosin binding protein C, slow-type |
Gene Aliases: |
8030451F13Rik |
Location: |
Chr.10:88518279-88605234 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mybpc1 | NM_001252372.1 | 11 | 1251 | NP_001239301.1 |
| NM_175418.5 | 12 | 1293 | NP_780627.2 | |
| AK132188.1 | 11 | 1124 | ||
| BC062158.1 | 12 | 1155 | AAH62158.1 |

Genomic Map