Assay Details
Target Gene Details
Entrez Gene ID: | 13713 |
Gene Name: | ELK3, member of ETS oncogene family |
Gene Aliases: |
D430049E23Rik, Erp, Etrp, Net, Sap-2 |
Location: |
Chr.10:93247415-93311167 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elk3 | NM_001282967.1 | NP_001269896.1 | ||
| NM_013508.2 | NP_038536.2 | |||
| AK052546.1 | BAC35034.1 | |||
| AK088129.1 | ||||
| AK133847.1 | BAE21883.1 | |||
| BC005686.1 | AAH05686.1 | |||
| BC053402.1 | AAH53402.1 | |||
| BC055735.1 | AAH55735.1 | |||
| FN434119.1 | ||||
| FN434120.1 | ||||
| FN434121.1 | ||||
| L19953.1 | AAA19872.1 | |||
| S82864.1 | AAB46893.1 | |||
| Z32815.1 | CAA83676.1 |
More Information
Set Membership: |
|

Genomic Map