Assay Details
Target Gene Details
Entrez Gene ID: | 70297 |
Gene Name: | GRIP and coiled-coil domain containing 2 |
Gene Aliases: |
0610043A03Rik, 2210420P05Rik, 2600014C01Rik, AW112121, mKIAA0336 |
Location: |
Chr.10:58254296-58305599 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcc2 | NM_027375.2 | 18 | 4195 | NP_081651.2 |
| AB093232.1 | 18 | 4198 | BAC41416.1 | |
| AK148283.1 | 13 | 1410 | BAE28456.1 | |
| BC065697.1 | 6 | 590 | AAH65697.1 | |
| BC139007.1 | 18 | 4191 |
More Information
Set Membership: |
|

Genomic Map