Assay Details
Target Gene Details
Entrez Gene ID: | 70297 |
Gene Name: | GRIP and coiled-coil domain containing 2 |
Gene Aliases: |
0610043A03Rik, 2210420P05Rik, 2600014C01Rik, AW112121, mKIAA0336 |
Location: |
Chr.10:58254296-58305599 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcc2 | NM_027375.2 | NP_081651.2 | ||
| AB093232.1 | BAC41416.1 | |||
| AK011206.1 | BAB27466.3 | |||
| AK045701.1 | BAC32463.2 | |||
| BC027339.1 | AAH27339.1 | |||
| BC139007.1 |
More Information
Set Membership: |
|

Genomic Map