Assay Details
Target Gene Details
Entrez Gene ID: | 11923 |
Gene Name: | neurogenic differentiation 4 |
Gene Aliases: |
AI846749, ATH-3, Atoh3, MATH-3, Math3, bHLHa4 |
Location: |
Chr.10:130268152-130280240 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neurod4 | NM_007501.4 | NP_031527.1 | ||
| AK012183.1 | BAB28083.1 | |||
| BC054391.1 | AAH54391.1 |
More Information
Set Membership: |
|

Genomic Map