Assay Details
Target Gene Details
Entrez Gene ID: | 11536 |
Gene Name: | G protein-coupled receptor 182 |
Gene Aliases: |
AM-R, Admr, G10-D, Gpcr17, Gpcr22, MB10, NOW |
Location: |
Chr.10:127749602-127751798 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr182 | NM_007412.2 | NP_031438.2 | ||
| BY121440.1 | ||||
| D17292.1 | BAA04125.1 |
More Information
Set Membership: |
|

Genomic Map