Assay Details
Target Gene Details
Entrez Gene ID: | 210198 |
Gene Name: | G protein-coupled receptor, family C, group 6, member A |
Gene Aliases: |
- |
Location: |
Chr.10:51614688-51631614 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gprc6a | NM_153071.1 | 1 | 149 | NP_694711.1 |
| AY101365.1 | 1 | 149 | AAM22280.1 | |
| AY735399.1 | 1 | 122 | AAW68012.1 | |
| AY735400.1 | 1 | 122 | AAW68013.1 | |
| AY735401.1 | 1 | 122 | AAW68014.1 | |
| BC108998.2 | 1 | 149 | AAI08999.1 |
More Information
Set Membership: |
|

Genomic Map