Assay Details
Target Gene Details
Entrez Gene ID: | 15109 |
Gene Name: | histidine ammonia lyase |
Gene Aliases: |
Hsd, his, histidase |
Location: |
Chr.10:93488768-93516743 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hal | NM_010401.3 | NP_034531.1 | ||
| AK014518.1 | BAB29407.1 | |||
| AK028963.1 | BAC26215.1 | |||
| AK081409.1 | ||||
| BC057637.1 | AAH57637.1 | |||
| BC157999.1 | ||||
| L07645.1 | AAA37777.1 |
More Information
Set Membership: |
|

Genomic Map