Assay Details
Target Gene Details
Entrez Gene ID: | 71912 |
Gene Name: | junctional sarcoplasmic reticulum protein 1 |
Gene Aliases: |
2300003C06Rik, 2310032K21Rik, JP-45, JP45 |
Location: |
Chr.10:80808494-80813593 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jsrp1 | NM_028001.3 | NP_082277.2 | ||
| AK009016.1 | ||||
| AK009578.1 | BAB26371.1 |

Genomic Map