Assay Details
Target Gene Details
Entrez Gene ID: | 66094 |
Gene Name: | LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae) |
Gene Aliases: |
0910001B06Rik, 1110033F18Rik |
Location: |
Chr.10:80852823-80855249 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsm7 | NM_025349.2 | NP_079625.1 | ||
| AK003062.1 | BAB22540.1 | |||
| AK007462.1 | BAB25051.1 | |||
| AK009317.1 | ||||
| AK012446.1 | BAB28245.1 | |||
| AK013355.1 | BAB28806.1 | |||
| BC027511.1 | AAH27511.1 | |||
| BC081444.1 | AAH81444.1 |
More Information
Set Membership: |
|

Genomic Map