Assay Details
Target Gene Details
Entrez Gene ID: | 56709 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 12 |
Gene Aliases: |
Dj10, mDj10 |
Location: |
Chr.10:59879569-59899296 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb12 | NM_019965.2 | NP_064349.2 | ||
| AB028860.1 | BAA88308.1 | |||
| AK038149.1 | ||||
| AK082546.1 | BAC38525.1 | |||
| AK089427.1 | ||||
| AK138495.1 | BAE23683.1 | |||
| AK166224.1 | BAE38642.1 | |||
| AK172274.1 | BAE42919.1 | |||
| AK172485.1 | ||||
| BC034162.1 | AAH34162.1 |

Genomic Map