Assay Details
Target Gene Details
Entrez Gene ID: | 73690 |
Gene Name: | GLI pathogenesis-related 1 (glioma) |
Gene Aliases: |
2410114O14Rik, RTVP-1, RTVP1, mRTVP-1 |
Location: |
Chr.10:111984792-112002619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glipr1 | NM_028608.3 | NP_082884.1 | ||
| AK010768.1 | BAB27168.1 | |||
| AK137557.1 | BAE23406.1 | |||
| AK170051.1 | BAE41532.1 | |||
| BC025083.1 | AAH25083.1 | |||
| BC098231.1 | AAH98231.1 |

Genomic Map