Assay Details
Target Gene Details
Entrez Gene ID: | 73690 |
Gene Name: | GLI pathogenesis-related 1 (glioma) |
Gene Aliases: |
2410114O14Rik, RTVP-1, RTVP1, mRTVP-1 |
Location: |
Chr.10:111984792-112002619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glipr1 | NM_028608.3 | NP_082884.1 | ||
| AK010768.1 | BAB27168.1 | |||
| AK137557.1 | BAE23406.1 | |||
| AK170051.1 | BAE41532.1 | |||
| BC025083.1 | AAH25083.1 | |||
| BC098231.1 | AAH98231.1 | |||
| BY540964.1 |
Target Gene Details
Entrez Gene ID: | 52705 |
Gene Name: | KRR1, small subunit (SSU) processome component, homolog (yeast) |
Gene Aliases: |
2610511F02Rik, AI255219, AI428520, D10Ertd773e, Hrb2 |
Location: |
Chr.10:111972695-111988430 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krr1 | NM_178610.4 | 10 | 2354 | NP_848725.2 |
| AK012108.1 | 10 | 2292 | ||
| AK030560.1 | 10 | 2266 | BAC27022.1 | |
| AK032472.1 | 10 | 2266 | BAC27888.1 | |
| AK036447.1 | 9 | 2352 | ||
| AK078627.1 | 4 | 1510 | BAC37343.1 | |
| AK146926.1 | 10 | 2271 | BAE27537.1 | |
| AK168675.1 | 10 | 2266 | BAE40525.1 | |
| BC089510.1 | AAH89510.1 | |||
| BC094236.1 | 10 | 2255 | AAH94236.1 |
More Information
Set Membership: |
|

Genomic Map