Assay Details
Target Gene Details
Entrez Gene ID: | 432467 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H3 |
Gene Aliases: |
AA693301, AI666703, Hnrph3 |
Location: |
Chr.10:63014650-63023889 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnph3 | NM_001079824.1 | 2 | 204 | NP_001073293.1 |
| BQ443117.1 | ||||
| BU946493.1 | 2 | 205 |
More Information
Set Membership: |
|

Genomic Map