Assay Details
Target Gene Details
Entrez Gene ID: | 432467 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H3 |
Gene Aliases: |
AA693301, AI666703, Hnrph3 |
Location: |
Chr.10:63014650-63023889 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnph3 | NM_001079824.1 | 4 | 543 | NP_001073293.1 |
| BQ443117.1 | 3 | 385 | ||
| BU946493.1 | 3 | 405 |
Target Gene Details
Entrez Gene ID: | 70432 |
Gene Name: | RUN and FYVE domain-containing 2 |
Gene Aliases: |
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13 |
Location: |
Chr.10:62980223-63018742 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rufy2 | NM_027425.3 | 18 | 8281 | NP_081701.2 |
| AK042856.1 | 1 | 1169 |
More Information
Set Membership: |
|

Genomic Map