Assay Details
Target Gene Details
Entrez Gene ID: | 30045 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 12 |
Gene Aliases: |
Jdp1, mJDP1 |
Location: |
Chr.10:63382443-63408840 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc12 | NM_001253685.1 | NP_001240614.1 | ||
| NM_013888.3 | NP_038916.1 | |||
| AF132906.2 | AAD48756.2 | |||
| AK002649.1 | BAB22258.1 | |||
| AK032207.1 | BAC27758.1 | |||
| BC013501.1 | AAH13501.1 | |||
| BF453706.1 | ||||
| DV042983.1 |
More Information
Set Membership: |
|

Genomic Map