Assay Details
Target Gene Details
Entrez Gene ID: | 17246 |
Gene Name: | transformed mouse 3T3 cell double minute 2 |
Gene Aliases: |
1700007J15Rik, AA415488, Mdm-2 |
Location: |
Chr.10:117688875-117710770 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mdm2 | NM_001288586.1 | NP_001275515.1 | ||
| NM_010786.4 | NP_034916.1 | |||
| AK004719.1 | BAB23502.1 | |||
| AK004881.1 | 7 | 1972 | ||
| AK088638.1 | BAC40470.1 | |||
| AK150530.1 | ||||
| AK152685.1 | ||||
| AK160069.1 | BAE35604.1 | |||
| AK168690.1 | ||||
| BC050902.1 | AAH50902.1 | |||
| BC092270.1 | AAH92270.1 | |||
| U47934.1 | AAB09031.1 | |||
| X58876.1 | CAA41684.1 |
More Information
Set Membership: |
|

Genomic Map