Assay Details
Target Gene Details
Entrez Gene ID: | 110829 |
Gene Name: | LIM and senescent cell antigen-like domains 1 |
Gene Aliases: |
2310016J22Rik, 4921524A02Rik, AI507642, AU021743, AW551584, C430041B13Rik, Lims1l, PINCH, PINCH1 |
Location: |
Chr.10:58323435-58424691 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lims1 | NM_001193303.1 | 2 | 304 | NP_001180232.1 |
| NM_026148.3 | 2 | 298 | NP_080424.2 | |
| NM_201242.1 | 2 | 322 | NP_957694.1 | |
| AK009390.1 | 2 | 305 | BAB26259.2 | |
| AK014954.1 | 2 | 299 | BAB29637.1 | |
| AK049564.1 | 1 | 638 | ||
| AK075667.1 | 1 | 71 | ||
| AK082932.1 | 2 | 167 | BAC38699.1 | |
| AK088939.1 | 2 | 173 | BAC40663.1 | |
| AK151235.1 | 2 | 173 | BAE30227.1 | |
| AK151938.1 | 2 | 165 | BAE30814.1 | |
| AK152679.1 | 2 | 165 | BAE31411.1 | |
| AK153346.1 | 2 | 173 | BAE31922.1 | |
| BC005621.1 | 2 | 153 | AAH05621.1 | |
| DQ003608.1 | 2 | 289 | AAY21820.1 | |
| EU122953.1 | 2 | 288 | ||
| EU122954.1 | 2 | 156 |
More Information
Set Membership: |
|

Genomic Map