Assay Details
Target Gene Details
Entrez Gene ID: | 100503659 |
Gene Name: | downstream of Stk11 |
Gene Aliases: |
9630008F14, R29144/1 |
Location: |
Chr.10:80130434-80140136 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dos | NM_001195268.1 | 8 | 1013 | NP_001182197.1 |
| NR_036582.1 | 10 | 1349 | ||
| AK035824.1 | 2 | 164 | BAC29200.1 | |
| AK137009.1 | 10 | 1349 | ||
| BC078442.1 | 4 | 568 | AAH78442.1 | |
| BC100552.1 | 3 | 396 | AAI00553.1 |
More Information
Set Membership: |
|

Genomic Map