Assay Details
Target Gene Details
Entrez Gene ID: | 15979 |
Gene Name: | interferon gamma receptor 1 |
Gene Aliases: |
CD119, IFN-gammaR, Ifgr, Ifngr, Nktar |
Location: |
Chr.10:19591958-19610225 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifngr1 | NM_010511.2 | NP_034641.1 | ||
| AF128216.1 | AAF22557.1 | |||
| AF128217.1 | AAF22558.1 | |||
| AK087525.1 | ||||
| AK150503.1 | BAE29616.1 | |||
| AK151079.1 | BAE30094.1 | |||
| AK152085.1 | BAE30935.1 | |||
| AK152384.1 | BAE31172.1 | |||
| AK153084.1 | BAE31709.1 | |||
| AK153347.1 | BAE31923.1 | |||
| AK159359.1 | BAE35017.1 | |||
| AK159660.1 | BAE35269.1 | |||
| AK159839.1 | BAE35417.1 | |||
| AK169593.1 | BAE41245.1 | |||
| AY157836.1 | AAO21311.1 | |||
| AY157837.1 | AAO21312.1 | |||
| BC132599.1 | ||||
| BC138617.1 | ||||
| J05265.1 | AAA39178.1 | |||
| M25764.1 | AAA39177.1 | |||
| M26711.1 | AAA37896.1 | |||
| M28233.1 | AAA37898.1 | |||
| M28995.1 | AAA37895.1 |
More Information
Set Membership: |
|

Genomic Map