Assay Details
Target Gene Details
Entrez Gene ID: | 114229 |
Gene Name: | KISS1 receptor |
Gene Aliases: |
Gpr54, KiSS-1 |
Location: |
Chr.10:79916971-79922282 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kiss1r | NM_053244.5 | 4 | 2282 | NP_444474.1 |
| AF343726.1 | 4 | 579 | AAK83236.1 | |
| AK039628.1 | 4 | 2283 | BAC30404.1 | |
| AK139303.1 | 2 | 2976 | ||
| AY044228.1 | 4 | 612 | AAK95814.1 | |
| BC016531.1 | 4 | 725 | AAH16531.1 | |
| EU532436.1 | ||||
| EU532437.1 | 4 | 579 | ||
| EU879091.1 | ||||
| GU075727.1 |
More Information
Set Membership: |
|

Genomic Map