Assay Details
Target Gene Details
Entrez Gene ID: | 108030 |
Gene Name: | lin-7 homolog A (C. elegans) |
Gene Aliases: |
AI848705, LIN-7A, MALS-1, TIP-33, Veli, Veli1 |
Location: |
Chr.10:107271798-107425146 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin7a | NM_001033223.2 | NP_001028395.1 | ||
| NM_001039354.1 | NP_001034443.1 | |||
| NM_001284329.1 | NP_001271258.1 | |||
| AK077191.1 | ||||
| AK160671.1 | BAE35951.1 | |||
| BC029721.1 | AAH29721.1 | |||
| BG288381.1 | ||||
| BU923246.1 | ||||
| BY123635.1 |
More Information
Set Membership: |
|

Genomic Map