Assay Details
Target Gene Details
Entrez Gene ID: | 17155 |
Gene Name: | mannosidase 1, alpha |
Gene Aliases: |
Man1a1, PCR1 |
Location: |
Chr.10:53904785-54076586 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Man1a | NM_008548.4 | 11 | 2197 | NP_032574.1 |
| AK031038.1 | 11 | 2198 | BAC27225.1 | |
| AK042371.1 | 10 | 2038 | ||
| AK043932.1 | 11 | 2138 | ||
| AK154820.1 | 11 | 2139 | BAE32852.1 | |
| AK171841.1 | 11 | 2140 | ||
| AK191835.1 | ||||
| AK192972.1 | ||||
| AK196316.1 | ||||
| AK196883.1 | 1 | 91 | ||
| AK197437.1 | 1 | 91 | ||
| AK201818.1 | 1 | 91 | ||
| AK204950.1 | ||||
| AK211829.1 | 1 | 91 | ||
| BC015265.1 | 11 | 2098 | AAH15265.1 | |
| U04299.1 | 10 | 1801 | AAA17747.1 |
More Information
Set Membership: |
|

Genomic Map